Praseodymium foil, 1.0mm (0.04in) thick, 99.5% (REO)
Praseodymium foil, 1.0mm (0.04in) thick, 99.5% (REO)
Praseodymium foil, 1.0mm (0.04in) thick, 99.5% (REO)
Thermo Scientific Chemicals

Praseodymium foil, 1.0mm (0.04in) thick, 99.5% (REO)

CAS: 7440-10-0 | Pr | 140.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 013963.FF
also known as 013963-FF
Price (EUR)
198,65
Online Exclusive
220,00
Save 21,35 (10%)
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (EUR)
198,65
Online Exclusive
220,00
Save 21,35 (10%)
Each
Chemical Identifiers
CAS7440-10-0
IUPAC Namepraseodymium
Molecular FormulaPr
InChI KeyPUDIUYLPXJFUGB-UHFFFAOYSA-N
SMILES[Pr]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.5% max.
Praseodymium is used as an alloying constituent for alloys used to make permanent magnets and flints. Along with neodymium, praseodymium is used to manufacture yellow glass which can be used as eye protection (e.g. for welders).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only