Neodymium foil, 0.1mm (0.004in) thick, 99.9% (REO)
Neodymium foil, 0.1mm (0.004in) thick, 99.9% (REO)
Neodymium foil, 0.1mm (0.004in) thick, 99.9% (REO)
Thermo Scientific Chemicals

Neodymium foil, 0.1mm (0.004in) thick, 99.9% (REO)

CAS: 7440-00-8 | Nd | 144.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 013964.FF
also known as 013964-FF
Price (EUR)
165,00
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (EUR)
165,00
Each
Chemical Identifiers
CAS7440-00-8
IUPAC Nameneodymium
Molecular FormulaNd
InChI KeyQEFYFXOXNSNQGX-UHFFFAOYSA-N
SMILES[Nd]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Neodymium applications include lasers, glass coloring and tinting, dielectrics and, most importantly, as the fundamental basis for neodymium-iron-boron (Nd2Fe14B) permanent magnets. It is also used in CRT displays to enhance contrast between reds and greens. It is highly valued in glass manufacturing for its attractive purple coloring to glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only