Barium sulfate, 97%
Barium sulfate, 97%
Barium sulfate, 97%
Thermo Scientific Chemicals

Barium sulfate, 97%

CAS: 7727-43-7 | BaO4S | 233.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
5 kg
Catalog number 013989.22
also known as 013989-22
Price (EUR)
36,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
36,00
Each
Chemical Identifiers
CAS7727-43-7
IUPAC Namebarium(2+) sulfate
Molecular FormulaBaO4S
InChI KeyTZCXTZWJZNENPQ-UHFFFAOYSA-L
SMILES[Ba++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size1-4 Microns average
Assay (unspecified)96.1% min
Barium sulfate has wide range of applications. It is primarily used in oil well drilling fluid, and as filler in the paint and varnish industry. It is one of the major constituents of the pigment Lithiopone and the white pigment for paints. It improves the resistance against acid and acidity for plastics when employed as filler. Further it is employed as a component in strobe pyrotechnic compositions, and as a coating for photographic papers to improve the effectiveness of the image. Baryta, a thin layer of barium sulfate, is used to make the papers bright for ink-jet printing. In the chemical and pharmaceutical industry, it is used as a catalyst support when selective hydrogenations are required, for e.g. in Rosenmund reduction (Pd on barium sulfate). In colorimetry, barium sulfate is used as a diffuser to measure light sources. In the medical field, barium sulfate suspension is used clinically as a radiocontrast agent for X-ray imaging and other diagnostic procedures. In biomaterials research, it is one of the components utilized in the preparation of acrylic bone cements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only