Dichloromethylsilane, 97+%
Dichloromethylsilane, 97+%
Dichloromethylsilane, 97+%
Thermo Scientific Chemicals

Dichloromethylsilane, 97+%

CAS: 75-54-7
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 014079.22
also known as 014079-22
Price (EUR)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS75-54-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (unspecified)≥97.0%
A monomer (‘building block’) in the production of silicone polymers. An intermediate (starting material) in the production of other organic and inorganic chemicals. In the electronics industry for the production of ultra-pure polysilicon and in the manufacture of semiconductors and photovoltaics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only