Tetra-n-butyltin, 95%
Tetra-n-butyltin, 95%
Tetra-n-butyltin, 95%
Thermo Scientific Chemicals

Tetra-n-butyltin, 95%

CAS: 1461-25-2 | C16H36Sn | 347.174 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 014115.22
also known as 014115-22
Price (EUR)
54,20
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
54,20
Each
Chemical Identifiers
CAS1461-25-2
IUPAC Nametetrabutylstannane
Molecular FormulaC16H36Sn
InChI KeyAFCAKJKUYFLYFK-UHFFFAOYSA-N
SMILESCCCC[Sn](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Pale yellow
Assay from Suppliers CofA≥94%
Tetra-n-butyltin is used in hydrogenolysis reactions and as a transition metal catalyst. It is also used stabilizers in the manufacture of polyvinyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only