Tin rod, 12.7mm (0.5in) dia, 99.9% (metals basis)
Tin rod, 12.7mm (0.5in) dia, 99.9% (metals basis)
Tin rod, 12.7mm (0.5in) dia, 99.9% (metals basis)
Thermo Scientific Chemicals

Tin rod, 12.7mm (0.5in) dia, 99.9% (metals basis)

CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
100 cm
Catalog number 014148.BU
also known as 014148-BU
Price (EUR)
124,00
Each
Quantity:
25 cm
Request bulk or custom format
Price (EUR)
124,00
Each
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
FormRod, 12.7mm (0.5in) diameter.
Assay from Supplier's CofA≥99.9% (metals basis)
Appearance (Color)Silvery
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only