Silver, Oil based standard solution, Specpure, Ag 1000
Silver, Oil based standard solution, Specpure, Ag 1000
Silver, Oil based standard solution, Specpure, Ag 1000
Thermo Scientific Chemicals

Silver, Oil based standard solution, Specpure, Ag 1000

 
Have Questions?
Catalog NumberQuantity
014199.18
also known as 014199-18
50 g
Catalog number 014199.18
also known as 014199-18
Price (EUR)
-
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSilver Oil based standard solution
MDL NumberMFCD00003397
Name NoteAg 1000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only