Manganese(II) carbonate, 99.9% (metals basis excluding Na)
Manganese(II) carbonate, 99.9% (metals basis excluding Na)
Manganese(II) carbonate, 99.9% (metals basis excluding Na)
Thermo Scientific Chemicals

Manganese(II) carbonate, 99.9% (metals basis excluding Na)

CAS: 598-62-9 | CMnO3 | 114.946 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 014344.A1
also known as 014344-A1
Price (EUR)
162,00
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
162,00
Each
Chemical Identifiers
CAS598-62-9
IUPAC Namemanganese(2+) carbonate
Molecular FormulaCMnO3
InChI KeyXMWCXZJXESXBBY-UHFFFAOYSA-L
SMILES[Mn++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentNa 1% max
Total Metal Impurities0.1% max.
Manganese(II) carbonate is used as an additive to plant fertilizers, glaze colorant in ceramics, pigment and drier for varnishes. It is also used as an ingredient in welding rod, animal feed additive as well as to prepare other manganese salts and pharmaceuticals. It is also used in medicine as a hematinic and in health foods, in flux and in concrete stains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only