Sodium, plasma standard solution, Specpure™ Na 10,000μg/mL
Thermo Scientific Chemicals

Sodium, plasma standard solution, Specpure™ Na 10,000μg/mL

Na2CO3 in 5% HNO3
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 014400.AE
also known as 014400-AE
Price (EUR)
144,00
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
144,00
Each
Specifications
Chemical Name or MaterialSodium, plasma standard solution
Concentration or Composition (by Analyte or Components)Nitric acid: 5%; Sodium carbonate: 2.3%; Water: 92.7%
DOT InformationDOT Class: 8, Packaging Group: III
EINECS Number231-714-2
Health Hazard 1H290-H314-H335
View more
Sodium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only