Aluminum, plasma standard solution, Specpure™ Al 10,000μg/mL
Thermo Scientific Chemicals

Aluminum, plasma standard solution, Specpure™ Al 10,000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 014405.AE
also known as 014405-AE
Price (EUR)
135,00
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
135,00
Each
Specifications
Chemical Name or MaterialAluminum plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: HYDROCHLORIC ACID
EINECS Number231-595-7
Health Hazard 1H290
Health Hazard 2GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.
View more
Aluminum, plasma standard solution is used as a single-element standard solution for plasma emission spectrometry. It is also used as a standard solution in analytical chemistry and atomic absorption spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only