Specifications
Chemical Name or MaterialBeryllium plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECS Number231-714-2
Health Hazard 1H290-H302-H314-H317-H331-H335-H350i-H373
Health Hazard 2GHS H Statement
H350-H372-H314-H302-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
View more