Silver, plasma standard solution, Specpure™ Ag 10,000μg/mL
Silver, plasma standard solution, Specpure™ Ag 10,000μg/mL
Silver, plasma standard solution, Specpure™ Ag 10,000μg/mL
Thermo Scientific Chemicals

Silver, plasma standard solution, Specpure™ Ag 10,000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 014418.AP
also known as 014418-AP
Price (EUR)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialSilver plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECS Number231-714-2
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only