Aluminum titanium oxide, 99.5% (metals basis)
Aluminum titanium oxide, 99.5% (metals basis)
Aluminum titanium oxide, 99.5% (metals basis)
Thermo Scientific Chemicals

Aluminum titanium oxide, 99.5% (metals basis)

CAS: 12004-39-6 | Al2O5Ti | 181.825 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 014484.14
also known as 014484-14
Price (EUR)
109,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
109,00
Each
Chemical Identifiers
CAS12004-39-6
IUPAC Namedititanium(4+) dialuminium(3+) heptaoxidandiide
Molecular FormulaAl2O7Ti2
InChI KeyCOTUMVFAJGQVPQ-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Al+3].[Al+3].[Ti+4].[Ti+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Aluminum titanium oxide is used in refractory industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only