Molybdenum wire, platinum coated, 0.457mm (0.018in) dia
Molybdenum wire, platinum coated, 0.457mm (0.018in) dia
Molybdenum wire, platinum coated, 0.457mm (0.018in) dia
Thermo Scientific Chemicals

Molybdenum wire, platinum coated, 0.457mm (0.018in) dia

CAS: 7439-98-7 | Mo | 95.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
1 m
5 m
Catalog number 014658.BU
also known as 014658-BU
Price (EUR)
-
Quantity:
25 cm
Request bulk or custom format
Specifications
CAS7439-98-7
ChEBICHEBI:28685
Chemical Name or MaterialMolybdenum wire, platinum coated
EINECS Number231-107-2
IUPAC Namemolybdenum
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only