Silver flake, 90% <20 micron, 99.9% (metals basis)
Silver flake, 90% <20 micron, 99.9% (metals basis)
Silver flake, 90% <20 micron, 99.9% (metals basis)
Thermo Scientific Chemicals

Silver flake, 90% <20 micron, 99.9% (metals basis)

Suitable for manufacturing metallizing preparations and conductive silicone rubbers | CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 x 10 g
Catalog number 014755.09
also known as 014755-09
Price (EUR)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max., 90% <20 micron
Suitable for manufacturing metallizing preparations and conductive silicone rubbers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only