Barium bromide, anhydrous, 99%, Thermo Scientific Chemicals
Barium bromide, anhydrous, 99%, Thermo Scientific Chemicals
Barium bromide, anhydrous, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium bromide, anhydrous, 99%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
016792.18
also known as 016792-18
50 g
Catalog number 016792.18
also known as 016792-18
Price (EUR)
81,90
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
81,90
Each
Chemical Identifiers
CAS10553-31-8
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
Barium bromide, anhydrous is used as the chemical intermediate, and the spice cosmetics chemical additive. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only