Bismuth(III) bromide, 99% (metals basis)
Bismuth(III) bromide, 99% (metals basis)
Bismuth(III) bromide, 99% (metals basis)
Thermo Scientific Chemicals

Bismuth(III) bromide, 99% (metals basis)

CAS: 7787-58-8 | BiBr3 | 448.692 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 017112.14
also known as 017112-14
Price (EUR)
131,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
131,00
Each
Chemical Identifiers
CAS7787-58-8
IUPAC Namebismuth(3+) tribromide
Molecular FormulaBiBr3
InChI KeyTXKAQZRUJUNDHI-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
It catalyzes the formation of cyclic carbonates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only