Calcium hydride, 90-95%
Calcium hydride, 90-95%
Calcium hydride, 90-95%
Thermo Scientific Chemicals

Calcium hydride, 90-95%

CAS: 7789-78-8 | CaH2
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 019106.14
also known as 019106-14
Price (EUR)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7789-78-8
IUPAC Namecalcium dihydride
Molecular FormulaCaH2
InChI KeyFAQLAUHZSGTTLN-UHFFFAOYSA-N
SMILES[CaH2]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to gray
FormGranules or powder or granular powder
Assay from Supplier's CofA≥88.0%
Calcium hydride is used as an efficient drying agent for aprotic base-stable solvents like ethers and tertiary amines. It is a useful dehydrating agent in a synthesis of aldehyde enamines in high yield and purity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only