Toluene, Spectrophotometric Grade, 99.7+%
Toluene, Spectrophotometric Grade, 99.7+%
Toluene, Spectrophotometric Grade, 99.7+%
Thermo Scientific Chemicals

Toluene, Spectrophotometric Grade, 99.7+%

Maximum level of impurities: Evaporation residue 4ppm, H2O 0.03% | CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 x 1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 019376.K2
also known as 019376-K2
Price (EUR)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS108-88-3
IUPAC Nametoluene
Molecular FormulaC7H8
InChI KeyYXFVVABEGXRONW-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
Color (APHA)10, max
Optical Absorbance at285nm, 1.00, max
Optical Absorbance at286nm, 0.70, max
Optical Absorbance at293nm, 0.20, max
View more
Toluene is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only