1,2,4-Trichlorobenzene, Spectrophotometric Grade, 99% min
1,2,4-Trichlorobenzene, Spectrophotometric Grade, 99% min
1,2,4-Trichlorobenzene, Spectrophotometric Grade, 99% min
Thermo Scientific Chemicals

1,2,4-Trichlorobenzene, Spectrophotometric Grade, 99% min

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.01% | CAS: 120-82-1 | C6H3Cl3 | 181.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 019390.K2
also known as 019390-K2
Price (EUR)
140,00
Each
Quantity:
1 L
Request bulk or custom format
Price (EUR)
140,00
Each
Chemical Identifiers
CAS120-82-1
IUPAC Name1,2,4-trichlorobenzene
Molecular FormulaC6H3Cl3
InChI KeyPBKONEOXTCPAFI-UHFFFAOYSA-N
SMILESClC1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
CommentCountry of origin:
Optical Absorbance at310nm: ≤0.5 AU
Residue on Evaporation≤5ppm
Assay (GC)≥99.0%
Optical Absorbance at308nm: ≤1 AU
View more
1,2,4-Trichlorobenzene is used as a dielectric and heat transfer fluid in transformers. It acts as an intermediate, degreaser, wood preservative and solvent for dye. It is a high-temperature solvent used in gel permeation chromatography, especially for polyethylene and polypropylene. Further, it is used as a lubricant and as a synthetic transformer oil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only