2-Propanol, Semiconductor Grade, 99.5% min
2-Propanol, Semiconductor Grade, 99.5% min
2-Propanol, Semiconductor Grade, 99.5% min
Thermo Scientific Chemicals

2-Propanol, Semiconductor Grade, 99.5% min

Maximum level of impurities: Color (APHA) 10, Evaporation residue 5ppm, Solubility in H2O P.T., H2O (by Karl Fischer titrn) 0.05%, Acidity 0.2meq/g, Alkalinity 0.1meq/g, Cl 0.2ppm, PO4 0.5ppm | CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 x 1 L
Catalog number 019397.M1
Price (EUR)
198,65
Online exclusive
220,00
Save 21,35 (10%)
Each
Quantity:
4 x 1 L
Request bulk or custom format
Price (EUR)
198,65
Online exclusive
220,00
Save 21,35 (10%)
Each
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentPO₄ 0.5ppm, max
Water Content (Karl Fischer Titration)0.05%, max
Residue on Evaporation5ppm, max
Solution Testin H₂O P.T.
FormClear liquid
View more
2-Propanol is used as a solvent for gums, resins, alkaloids and essential oils, as a denaturant for ethyl alcohol and is used in lotions and cosmetics. It is used in keyboard, LCD and laptop cleaning, as a wetting agent in the fountain solution used in lithographic printing, and often used as a solvent for French polishing shellac used in cabinet making. It acts as hydride source in the Meerwein-Ponndorf-Verley reduction. It plays an important role in DNA extraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only