1-n-Butylpyridinium chloride, 98%
1-n-Butylpyridinium chloride, 98%
1-n-Butylpyridinium chloride, 98%
1-n-Butylpyridinium chloride, 98%
Thermo Scientific Chemicals

1-n-Butylpyridinium chloride, 98%

CAS: 1124-64-7 | C9H14ClN | 171.668 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 019740.04
also known as 019740-04
Price (EUR)
53,90
Each
Quantity:
2 g
Request bulk or custom format
Price (EUR)
53,90
Each
Chemical Identifiers
CAS1124-64-7
IUPAC Name1-butylpyridin-1-ium
Molecular FormulaC9H14N
InChI KeyREACWASHYHDPSQ-UHFFFAOYSA-N
SMILESCCCC[N+]1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormPowder or crystalline powder or solid
Assay (HPLC)≥97.5%
Proton NMRConforms to structure
The derivatives of 1-n-Butylpyridinium chloride is used in spent fuel reprocessing, particularly in the extraction of high-level radioactive aqueous waste from the processing of nuclear fuel. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only