Tin(II) ethoxide
Tin(II) ethoxide
Tin(II) ethoxide
Thermo Scientific Chemicals

Tin(II) ethoxide

CAS: 14791-99-2 | C4H10O2Sn | 208.832 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 020896.03
also known as 020896-03
Price (EUR)
47,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
47,00
Each
Chemical Identifiers
CAS14791-99-2
IUPAC Nameλ²-tin(2+) diethanolate
Molecular FormulaC4H10O2Sn
InChI KeyXCKWFNSALCEAPW-UHFFFAOYSA-N
SMILES[Sn++].CC[O-].CC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay from Supplier's CofATin (Sn): Report %
FormPowder
Tin(II) ethoxide is used in catalyst synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only