Cerium(III) oxalate hydrate, REacton™, 99.9% (REO)
Cerium(III) oxalate hydrate, REacton™, 99.9% (REO)
Cerium(III) oxalate hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Cerium(III) oxalate hydrate, REacton™, 99.9% (REO)

CAS: 15750-47-7 | C6Ce2O12 | 544.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 021122.14
also known as 021122-14
Price (EUR)
89,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
89,00
Each
Chemical Identifiers
CAS15750-47-7
IUPAC Namedicerium(3+) trioxalate
Molecular FormulaC6Ce2O12
InChI KeyZMZNLKYXLARXFY-UHFFFAOYSA-H
SMILES[Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only