Calcium aluminum oxide, 99% (metals basis)
Calcium aluminum oxide, 99% (metals basis)
Calcium aluminum oxide, 99% (metals basis)
Thermo Scientific Chemicals

Calcium aluminum oxide, 99% (metals basis)

CAS: 12042-68-1 | Al2CaO4 | 158.037 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 022646.14
also known as 022646-14
Price (EUR)
171,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
171,00
Each
Chemical Identifiers
CAS12042-68-1
IUPAC Namecalcium dialumanoylolate
Molecular FormulaAl2CaO4
InChI KeyXFWJKVMFIVXPKK-UHFFFAOYSA-N
SMILES[Ca++].[O-][Al]=O.[O-][Al]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Calcium aluminum oxide is used as a precursor in the production of naphtha steam reforming catalysts, refinery gas steam reforming catalysts, aluminum chlorohydrate, secondary hydrocarbon steam reforming catalysts. It is also used in the production of refractory and cements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only