Nickel(II) iodide, anhydrous, 99.5% (metals basis)
Nickel(II) iodide, anhydrous, 99.5% (metals basis)
Nickel(II) iodide, anhydrous, 99.5% (metals basis)
Nickel(II) iodide, anhydrous, 99.5% (metals basis)
Thermo Scientific Chemicals

Nickel(II) iodide, anhydrous, 99.5% (metals basis)

CAS: 13462-90-3 | I2Ni | 312.502 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 022893.14
also known as 022893-14
Price (EUR)
198,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
198,00
Each
Add to cart
Chemical Identifiers
CAS13462-90-3
IUPAC Namenickel(2+) diiodide
Molecular FormulaI2Ni
InChI KeyBFSQJYRFLQUZKX-UHFFFAOYSA-L
SMILES[Ni++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Nickel(II) iodide serves as a catalyst in carbonylation reactions. It is also used as a reagent in organic synthesis, especially in conjunction with samarium(II) iodide. It plays an important role in homogeneous catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only