1,4-Dioxane, HPLC Grade, 99% min
1,4-Dioxane, HPLC Grade, 99% min
1,4-Dioxane, HPLC Grade, 99% min
Thermo Scientific Chemicals

1,4-Dioxane, HPLC Grade, 99% min

CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 x 1 L
Catalog number 022913.K2
also known as 022913-K2
Price (EUR)
178,00
Each
Quantity:
1 L
Request bulk or custom format
Price (EUR)
178,00
Each
Chemical Identifiers
CAS123-91-1
IUPAC Name1,4-dioxane
Molecular FormulaC4H8O2
InChI KeyRYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILESC1COCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water0.03%
Residue on Evaporation5ppm
1,4-Dioxane HPLC Grade is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only