Zirconium(IV) n-propoxide, 70% w/w in n-propanol, Thermo Scientific Chemicals
Zirconium(IV) n-propoxide, 70% w/w in n-propanol, Thermo Scientific Chemicals
Zirconium(IV) n-propoxide, 70% w/w in n-propanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium(IV) n-propoxide, 70% w/w in n-propanol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
500 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 022989.36
Price (EUR)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS23519-77-9
IUPAC Nametetrapropoxyzirconium
Molecular FormulaC12H28O4Zr
InChI KeyXPGAWFIWCWKDDL-UHFFFAOYSA-N
SMILESCCCO[Zr](OCCC)(OCCC)OCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
FormLiquid
Assay (unspecified)ZrO2: 27.0 to 29.0%
Chloride content≤500 ppm
Specific Gravity1.05-1.09
Zirconium(IV) n-propoxide is used as a precursor in the production of zirconia. It is also used to study the thermophysical properties like thermal conductivity and thermal effusivity of hybrid films and thereby adjusting optic properties of the films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only