Tricarbonylnitrosylcobalt
Tricarbonylnitrosylcobalt
Tricarbonylnitrosylcobalt
Thermo Scientific Chemicals

Tricarbonylnitrosylcobalt

CAS: 14096-82-3
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 023151.03
also known as 023151-03
Price (EUR)
71,40
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
71,40
Each
Specifications
CAS14096-82-3
Chemical Name or MaterialTricarbonylnitrosylcobalt
ColorRed
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
Density1.47 g/mL
View more
Tricarbonylnitrosylcobalt is used as the Co metalorganic precursor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only