Tetra-n-hexylammonium perchlorate
Tetra-n-hexylammonium perchlorate
Tetra-n-hexylammonium perchlorate
Thermo Scientific Chemicals

Tetra-n-hexylammonium perchlorate

CAS: 4656-81-9 | C24H52ClNO4 | 454.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 030830.06
also known as 030830-06
Price (EUR)
43,20
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
43,20
Each
Specifications
CAS4656-81-9
Chemical Name or MaterialTetra-n-hexylammonium perchlorate
DOT InformationTransport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECS Number225-095-8
Formula Weight454.13
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only