1,5-Diphenylcarbazide, ACS
1,5-Diphenylcarbazide, ACS
1,5-Diphenylcarbazide, ACS
Thermo Scientific Chemicals

1,5-Diphenylcarbazide, ACS

CAS: 140-22-7 | C13H14N4O | 242.282 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 031390.06
also known as 031390-06
Price (EUR)
30,60
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
30,60
Each
Chemical Identifiers
CAS140-22-7
IUPAC Name1,3-bis(phenylamino)urea
Molecular FormulaC13H14N4O
InChI KeyKSPIHGBHKVISFI-UHFFFAOYSA-N
SMILESO=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream; slowly becomes pink on exposure to air.
FormCrystalline powder or powder
Assay (HPLC)≥97.0%
Identification (FTIR)Conforms
Residue on ignition≤0.05%
View more
1,5-Diphenylcarbazide is used in the colorimetric determination of chromium, osmium as well as in the detection of cadmium, mercury, magnesium, emetine and aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only