1,3-Diphenylguanidine, primary standard, 99+%
1,3-Diphenylguanidine, primary standard, 99+%
1,3-Diphenylguanidine, primary standard, 99+%
Thermo Scientific™

1,3-Diphenylguanidine, primary standard, 99+%

Primary material for standardizing acids | CAS: 102-06-7 | C13H13N3 | 211.268 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 031395.18
also known as 031395-18
Price (EUR)
77,50
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
77,50
Each
Specifications
Beilstein1875653
CAS102-06-7
Chemical Name or Material1,3-Diphenylguanidine
ColorCream
DOT InformationTransport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
View more
Primary material for standardizing acids. Diphenylguanidine is used as a medium of natural and synthetic rubber accelerator. It acts as a complexing agent for the detection of metals and organic bases. Further, it is used as a secondary foam stabilizer in the silicoflouride foam process. It is actively involved in the manufacture of rubber sheet, rubber shoes and other rubber industrial products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only