Boron phosphate
Boron phosphate
Boron phosphate
Thermo Scientific Chemicals

Boron phosphate

CAS: 13308-51-5 | BO4P | 105.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 032562.36
also known as 032562-36
Price (EUR)
605,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
605,00
Each
Chemical Identifiers
CAS13308-51-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay from Supplier's CofA≥96.0%
FormPowder
Boron phosphate is used as a catalyst for dehydration as well as in other organic reactions. It is employed as a source of phosphates involved in the exchange reaction to prepare metal phosphates. It finds application in refractories, ceramics and flame-retardants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only