Al-23 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 40 x 30 x 26;Vol (ml), 20
Thermo Scientific Chemicals

Al-23 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 40 x 30 x 26;Vol (ml), 20

Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) OuterDiameter (Metric) InnerHeight (Metric)
032951.KF
also known as 032951-KF
6 mL20mm16mm30mm
032955.KF
also known as 032955-KF
110 mL50mm44mm75mm
032953.KF
also known as 032953-KF
40 mL35mm30mm50mm
032954.KF
also known as 032954-KF
60 mL40mm36mm60mm
032952.KF
also known as 032952-KF
20 mL30mm26mm40mm
Catalog number 032951.KF
also known as 032951-KF
Price (EUR)
66,30
Each
Capacity (Metric):
6 mL
Diameter (Metric) Outer:
20mm
Diameter (Metric) Inner:
16mm
Height (Metric):
30mm
Price (EUR)
66,30
Each
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Specifications
Diameter (Metric) Inner16mm
Diameter (Metric) Outer20mm
Height (Metric)30mm
Quantity1 Piece
Capacity (Metric)6 mL
MaterialAl-23
Product TypeCylindrical Al-23 Crucibles
Unit SizeEach

Frequently asked questions (FAQs)

What is the maximum working temperature of Al-23 Tubes?

The maximum working temperature for the Al-23 Tube is 1950 degrees C.