Hydrogen peroxide, ACS, 29-32% w/w aq. soln., stab.
Hydrogen peroxide, ACS, 29-32% w/w aq. soln., stab.
Hydrogen peroxide, ACS, 29-32% w/w aq. soln., stab.
Thermo Scientific Chemicals

Hydrogen peroxide, ACS, 29-32% w/w aq. soln., stab.

For Bleaching and Deodorizing Textiles, Wood, Hair | CAS: 7722-84-1 | H2O2 | 34.014 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
500 mL
4 x 500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 033323.AP
also known as 033323-AP
Price (EUR)
55,65
Online Exclusive
61,80
Save 6,15 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (EUR)
55,65
Online Exclusive
61,80
Save 6,15 (10%)
Each
Chemical Identifiers
CAS7722-84-1
IUPAC Nameperoxol
Molecular FormulaH2O2
InChI KeyMHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILESOO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear
Color (APHA)≤10
Assay (Titration)29.0-32.0% H2O2
Chloride content≤3ppm
Free acid (titration)≤0.0006 meq/g
View more
For bleaching and deodorizing textiles, wood, hair, etc; rocket propulsion; maturing agent in food; water and sewage treatment; disinfectant; laboratory reagentHydrogen peroxide is used as an oxidizing agent to prepare propylene oxide, dibenzoyl peroxide, lauryl peroxide, peracetic acid and meta-chloroperoxybenzoic acid. It is also involved in the production of inorganic chemicals like sodium perborate and sodium percarbonate, which are used as bleaching agents in detergents. It is an associated iron catalyst and utilized for waste-water treatment to remove organic impurities through an advanced oxidation process. It is also used as a laboratory disinfectant and as a topical antiseptic. In the cosmetic industry, it is used for bleaching human hair and tooth whitening by mixing with ammonium hydroxide and baking soda respectively. Monopropellant engines using hydrogen peroxide as the fuel are commonly used for air-independent underwater propulsion applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only