Magnesium sulfate, anhydrous, 99.5% min
Magnesium sulfate, anhydrous, 99.5% min
Magnesium sulfate, anhydrous, 99.5% min
Thermo Scientific Chemicals

Magnesium sulfate, anhydrous, 99.5% min

CAS: 7487-88-9 | MgO4S | 120.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
10 kg
Catalog number 033337.36
also known as 033337-36
Price (EUR)
67,20
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
67,20
Each
Chemical Identifiers
CAS7487-88-9
IUPAC Namemagnesium(2+) sulfate
Molecular FormulaMgO4S
InChI KeyCSNNHWWHGAXBCP-UHFFFAOYSA-L
SMILES[Mg++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying2% max
Trace Metals0.001% max
Impurity content0.003% Se max
FormPowder
Assay (unspecified)99.5% min
View more
Magnesium sulfate is suitable for sensitive molecular biology applications. It used as a component of bath salts and beauty product. It is utilized by the athletes to soothe sore muscles and effective in the removal of splinters. It is commonly used as a desiccant in organic synthesis due to its affinity for water. It acts as a brewing salt in beer production to adjust the ion content of the brewing water as well as enhance enzyme action. It plays a vital role as an antidote for barium chloride poisoning. . It is useful in controlling seizures associated with epilepsy, glomerulonephritis, or hypothyroidism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only