Sodium thiocyanate, ACS, 98.0% min
Sodium thiocyanate, ACS, 98.0% min
Sodium thiocyanate, ACS, 98.0% min
Thermo Scientific Chemicals

Sodium thiocyanate, ACS, 98.0% min

CAS: 540-72-7 | CNNaS | 81.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 033388.22
also known as 033388-22
Price (EUR)
38,90
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
38,90
Each
Chemical Identifiers
CAS540-72-7
IUPAC Namesodium cyanosulfanide
Molecular FormulaCNNaS
InChI KeyVGTPCRGMBIAPIM-UHFFFAOYSA-M
SMILES[Na+].[S-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.0% NaSCN min
Impurity contentFe 2ppm
Impurity contentNH₄ 0.002%
Impurity contentS 0.001%
Impurity contentSO₄ 0.01%
View more

Sodium thiocyanate is used as a precursor for the synthesis of pharmaceuticals and specialty chemicals. It is used to prepare alkylthiocyanates by reacting with alkyl halides. It is involved nucleophilic substitution reactions with phase-transfer catalysts. It is a common laboratory reagent used to test presence of Fe3+ ions. It acts as a spinning solvent for polyacrylonitrile in fiber industry, photographic stabilizer, corrosion inhibitor, polymer stabilizer and hardening accelerator for cement and concrete mixes. Further, it is also used in both the crystallization and purification of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only