Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct
Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct
Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct
Thermo Scientific Chemicals

Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct

CAS: 75965-35-4 | C36H50BLiO6 | 596.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 035445.02
also known as 035445-02
Price (EUR)
43,50
Each
Quantity:
0.5 g
Request bulk or custom format
Price (EUR)
43,50
Each
Chemical Identifiers
CAS75965-35-4
IUPAC Namelithium(1+) tris(1,2-dimethoxyethane) tetraphenylboranuide
Molecular FormulaC36H50BLiO6
InChI KeyADYUZFWVPWDPFK-UHFFFAOYSA-N
SMILES[Li+].COCCOC.COCCOC.COCCOC.C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥98.0%
Appearance (Color)White to off-white
FormPowder or crystals or crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only