Nitrate, Ion chromatography standard solution, Specpure™ NO{3}∣- 1000μg/mL
Thermo Scientific Chemicals

Nitrate, Ion chromatography standard solution, Specpure™ NO{3}∣- 1000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 035549.AE
also known as 035549-AE
Price (EUR)
129,65
Online Exclusive
144,00
Save 14,35 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
129,65
Online Exclusive
144,00
Save 14,35 (10%)
Each
Specifications
Chemical Name or MaterialNitrate Ion chromatography standard solution
MDL NumberMFCD00144920
Name NoteNO3- 1000μg/mL
Physical FormLiquid
Quantity100 mL
View more
Nitrate, Ion chromatography standard solution is used as a standard solution in analytical chemistry and in ion chromatography. It is used for calibration of ion chromatography and other analytical techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only