Potassium hydroxide, 0.5N Standardized Solution
Potassium hydroxide, 0.5N Standardized Solution
Potassium hydroxide, 0.5N Standardized Solution
Thermo Scientific Chemicals

Potassium hydroxide, 0.5N Standardized Solution

CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
20 L
Catalog number 035595.K2
also known as 035595-K2
Price (EUR)
35,00
Each
Quantity:
1 L
Request bulk or custom format
Price (EUR)
35,00
Each
Chemical Identifiers
CAS1310-58-3
IUPAC Namepotassium hydroxide
Molecular FormulaHKO
InChI KeyKWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES[OH-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Concentration0.500N ± 0.14% (0.0014N)
Potassium hydroxide, 0.5N Standardized Solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only