Potassium hydrogen iodate, 0.025N Standardized Solution
Potassium hydrogen iodate, 0.025N Standardized Solution
Potassium hydrogen iodate, 0.025N Standardized Solution
Thermo Scientific Chemicals

Potassium hydrogen iodate, 0.025N Standardized Solution

CAS: 13455-24-8 | HI2KO6 | 389.909 g/mol
Have Questions?
Catalog NumberQuantity
035600.K2
also known as 035600-K2
1 L
Catalog number 035600.K2
also known as 035600-K2
Price (EUR)
134,00
Each
Quantity:
1 L
Request bulk or custom format
Price (EUR)
134,00
Each
Chemical Identifiers
CAS13455-24-8
IUPAC Namepotassium iodic acid iodate
Molecular FormulaHI2KO6
InChI KeyACAYDTMSDROWHW-UHFFFAOYSA-M
SMILES[K+].O[I](=O)=O.[O-][I](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration0.025N
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only