Sodium carbonate, 0.05N Standardized Solution
Sodium carbonate, 0.05N Standardized Solution
Sodium carbonate, 0.05N Standardized Solution
Thermo Scientific Chemicals

Sodium carbonate, 0.05N Standardized Solution

CAS: 497-19-8 | CNa2O3 | 105.99 g/mol
Have Questions?
Catalog NumberQuantity
035609.K2
also known as 035609-K2
1 L
Catalog number 035609.K2
also known as 035609-K2
Price (EUR)
39,10
Each
Quantity:
1 L
Request bulk or custom format
Price (EUR)
39,10
Each
Chemical Identifiers
CAS497-19-8
IUPAC Namedisodium carbonate
Molecular FormulaCNa2O3
InChI KeyCDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
FormLiquid
Assay from Supplier's CofA0.0499 N - 0.0501 N
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only