Lead(II) bromide, ultra dry, 99.999% (metals basis)
Lead(II) bromide, ultra dry, 99.999% (metals basis)
Lead(II) bromide, ultra dry, 99.999% (metals basis)
Thermo Scientific Chemicals

Lead(II) bromide, ultra dry, 99.999% (metals basis)

CAS: 10031-22-8 | Br2Pb | 367.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 035703.14
Price (EUR)
908,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
908,00
Each
Chemical Identifiers
CAS10031-22-8
IUPAC Nameλ²-lead(2+) dibromide
Molecular FormulaBr2Pb
InChI KeyZASWJUOMEGBQCQ-UHFFFAOYSA-L
SMILES[Br-].[Br-].[Pb++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey
Residual water≤100 ppm
Total Metal Impurities≤0.001%
FormBeads
Lead(II) bromide is used in the field of antirust, pigment and photograph. The molten lead(II) bromide acts as an electrolyte. Lead(II) bromide provides a high concentration of lead(II) ions and bromide ions to carry the current during the electrolysis process. The rare-earth-doped alkali-lead bromide crystals (potassium lead bromide (or) KPB; rubidium lead bromide or RPB) emerge as promising new low-phonon-energy host materials for mid-IR applications and are useful for solid state lasers. Hybrid organic/lead halide perovskites are promising materials for solar cell fabrication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only