Silver chloride, ultra dry, 99.997% (metals basis)
Silver chloride, ultra dry, 99.997% (metals basis)
Silver chloride, ultra dry, 99.997% (metals basis)
Thermo Scientific Chemicals

Silver chloride, ultra dry, 99.997% (metals basis)

CAS: 7783-90-6 | AgCl | 143.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 035715.06
also known as 035715-06
Price (EUR)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7783-90-6
IUPAC Namesilver(1+) chloride
Molecular FormulaAgCl
InChI KeyHKZLPVFGJNLROG-UHFFFAOYSA-M
SMILES[Cl-].[Ag+]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-10 Mesh
Appearance (Color)White to tan
FormBead
Total Metal Impurities0.003% max.
It is used in silver plating, electrode, photographic paper, in making antiseptic silver preparations. It is used to make photographic film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only