Ammonium thiocyanate, ACS, 97.5% min
Ammonium thiocyanate, ACS, 97.5% min
Ammonium thiocyanate, ACS, 97.5% min
Thermo Scientific Chemicals

Ammonium thiocyanate, ACS, 97.5% min

Fertilizers, photography, fabric dyeing, zinc coating, electroplating, and as a separator of Zr and Hf | CAS: 1762-95-4 | CH4N2S | 76.117 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 036247.30
also known as 036247-30
Price (EUR)
66,80
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
66,80
Each
Chemical Identifiers
CAS1762-95-4
IUPAC Nameammonium cyanosulfanide
Molecular FormulaCH4N2S
InChI KeySOIFLUNRINLCBN-UHFFFAOYSA-N
SMILES[NH4+].[S-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentIodine-consuming substances 0.004 meq/g
Trace Metals(as Pb) 5ppm
Impurity contentFe 3ppm
Impurity contentSO₄ 0.005%
pHof a 5% solution 4.5-6.0 at 25?
View more
Ammonium thiocyanate has many uses in manufacturing chemicals such as metal thiocyanates [CuSCN, Ca(SCN)2, NaSCN, KSCN], and in a number of pharmaceuticals. It is used in freezing solutions, pickling, printing, and corrosion inhibitor against acid gases. It is used in photography industry as a stabilizer or accelerator. Used in fertilizers, fabric dyeing, zinc coating, electroplating, and as a separator of Zr and Hf, also Au and Fe.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only