Gold Tin eutectic foil, 0.1mm (0.004 in.) thick x 43.8mm (1.72in) wide, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold Tin eutectic foil, 0.1mm (0.004 in.) thick x 43.8mm (1.72in) wide, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
036310.BS
also known as 036310-BS
10 cm
Catalog number 036310.BS
also known as 036310-BS
Price (EUR)
1.452,00
Each
Quantity:
10 cm
Request bulk or custom format
Price (EUR)
1.452,00
Each
Specifications
Assay Percent Notes(metals basis)
Avg. Mol. Wt. or Mol. Wt. RangeAu:Sn; 80:20 wt%
Chemical Name or MaterialGold Tin eutectic foil
MDL NumberMFCD02091709
Name Note0.1mm (0.004 in.) thick x 43.8mm (1.72 in.) wide
View more
Gold Tin eutectic foil is used in electronic manufacturing. It can be made into dimensions much smaller than soft solders, thereby it is utilized in applications which require the highest level of precision.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only