Lactic acid, ACS, 85.0-90.0% aq. soln.
Lactic acid, ACS, 85.0-90.0% aq. soln.
Lactic acid, ACS, 85.0-90.0% aq. soln.
Lactic acid, ACS, 85.0-90.0% aq. soln.
Thermo Scientific Chemicals

Lactic acid, ACS, 85.0-90.0% aq. soln.

CAS: 50-21-5 | C3H6O3 | 90.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 036415.36
also known as 036415-36
Price (EUR)
69,30
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (EUR)
69,30
Each
Add to cart
Chemical Identifiers
CAS50-21-5
IUPAC Name2-hydroxypropanoic acid
Molecular FormulaC3H6O3
InChI KeyJVTAAEKCZFNVCJ-UHFFFAOYNA-N
SMILESCC(O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormSyrupy liquid
Assay from Suppliers CofA85.0-90.0%
Chloride content≤0.001%
Residue on ignition≤0.02%
View more
It is used most commonly by athletes to describe the intense pain felt during exhaustive exercise. Used to make cultured dairy products, as a food preservative, and to make chemicals. Useful for the creation of highly aligned electrospun poly-L-lactic acid fibers for nerve regeneration applications. Poly(lactic acid) nanocomposites with various organoclays shows useful application for gas permeability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only