Dimethyl sulfoxide, ACS, 99.9% min
Dimethyl sulfoxide, ACS, 99.9% min
Dimethyl sulfoxide, ACS, 99.9% min
Thermo Scientific Chemicals

Dimethyl sulfoxide, ACS, 99.9% min

CAS: 67-68-5 | C2H6OS | 78.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 x 1 L
Catalog number 036480.AP
also known as 036480-AP
Price (EUR)
52,65
Online Exclusive
58,60
Save 5,95 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (EUR)
52,65
Online Exclusive
58,60
Save 5,95 (10%)
Each
Chemical Identifiers
CAS67-68-5
IUPAC Namemethanesulfinylmethane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILESCS(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water≤0.1%
FormLiquid
Free acid (titration)≤0.039 mg/g (≤0.001 meq/g)
Appearance (Color)Clear, colorless
Assay from Supplier's CofA≥99.9%
View more
It is used as a solvent for chemical reactions involving salts, most notably in Finkelstein reactions and in other nucleophilic substitutions. Used in antifreeze or hydraulic fluids. Utilized in the oxidation of thiols and disulfides to sulfonic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only