Calcium 2-ethylhexanoate, Superconductor Grade, in 2-ethylhexanoic acid (3-8% Ca)
Calcium 2-ethylhexanoate, Superconductor Grade, in 2-ethylhexanoic acid (3-8% Ca)
Calcium 2-ethylhexanoate, Superconductor Grade, in 2-ethylhexanoic acid (3-8% Ca)
Thermo Scientific Chemicals

Calcium 2-ethylhexanoate, Superconductor Grade, in 2-ethylhexanoic acid (3-8% Ca)

CAS: 136-51-6 | C16H30CaO4 | 326.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 036657.06
also known as 036657-06
Price (EUR)
40,90
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
40,90
Each
Specifications
CAS136-51-6
Chemical Name or MaterialCalcium 2-ethylhexanoate
Concentration or Composition (by Analyte or Components)3 to 8% Ca
EINECS Number205-249-0
Formula Weight326.5
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only