Thermo Scientific Chemicals

Methyl Red, ACS

Catalog number: 036682.22
100 g, Each
Thermo Scientific Chemicals

Methyl Red, ACS

Catalog number: 036682.22
100 g, Each
Quantity
Catalog number: 036682.22
also known as 036682-22
Price (EUR)
Price: 73,50
Online price: 66,15
Your price:
Quantity
-

Chemical Identifiers

CAS
493-52-7
IUPAC Name
2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid
Molecular Formula
C15H15N3O2
InChI Key
CEQFOVLGLXCDCX-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O
Comment
Clarity of alcohol solution P.T.
Comment
Visual transition interval from pH 4.2 (pink) to 6.2 (yellow)
Melting Point
(range) 179-182?

Description

pH indicator in pH range 4.2 - 6.3Methyl red is used as a pH indicator. It is red in pH less than 4.4 and yellow in pH over 6.2. It is used to study its degradation using Fenton's reagent. In microbiology, it is used to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Not Hazardous

    Not Hazardous